Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin A + C R482W Antibody (5H8-B4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$204.00 - $482.50
Specifications
Antigen | Lamin A + C R482W |
---|---|
Clone | 5H8-B4 |
Dilution | Western Blot 1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP177402SS
|
Novus Biologicals
NBP177402SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP177402
|
Novus Biologicals
NBP177402 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
Lamin A + C R482W Monoclonal specifically detects Lamin A + C R482W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Lamin A + C R482W | |
Western Blot 1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
NP_733821.1 | |
4000 | |
Hepatitis B Virus Capsid Antigen (HBcAg) Lamin A + C Ig-fold R482W fusion protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5H8-B4 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cancer, Chromatin Modifiers, Chromatin Research, DNA replication Transcription Translation and Splicing, Nuclear Envelope Markers, Signal Transduction | |
Ascites with 0.1% Sodium Azide | |
70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R82W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type) | |
LMNA | |
IgG1 κ | |
Unpurified | |
Disease-associated point-mutant R482W of Lamin A + C |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title