Learn More
anti-Lamin C, Clone: EM-11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP150051
Catalog No. NBP150051
Add to cart
Description
Description
Lamin A + C Monoclonal antibody specifically detects Lamin A + C in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
Lamin C
EM-11
Unconjugated
PBS (pH 7.4) with 15mM Sodium Azide
CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32
Mouse
IgG2a
0.1 mg
Store at 4C. Do not freeze.
Monoclonal
The mouse monoclonal antibody EM-11 recognizes lamin C, intermediate filament protein of nuclear lamina.
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:50
Purified
LMNA
The Lamin C antibody recognizes lamin C an intermediate filament protein of the nuclear lamina.
Protein A purified
RUO
Primary
4000
Human, Mouse