Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LDLR Mouse anti-Human, Alexa Fluor 488, Clone: 11A2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222035AF488
Description
LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
LDLR | |
Monoclonal | |
Alexa Fluor 488 | |
P01130 | |
LDLR | |
Human recombinant LDLR | |
0.1 mL | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
3949 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA | |
11A2 | |
Western Blot, Flow Cytometry, ELISA | |
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only