Learn More
LDLR Mouse anti-Human, Rat, Porcine, Bovine, Biotin, Clone: C7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178159B
Catalog No. NBP178159B
Add to cart
Description
Description
LDLR Monoclonal antibody specifically detects LDLR in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
LDLR
C7
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
P01130
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Mouse
IgG2b
0.1 ml
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
3949
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Biotin
PBS with 0.05% Sodium Azide
Purified
LDLR
Partially purified bovine LDL receptor [Swiss-Prot# P01131].
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Bovine, Human, Porcine, Rat