Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LIN-28A, Clone: 6D1F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128865

Catalog No. NBP128865


Add to cart

Description

Description

LIN-28A Monoclonal antibody specifically detects LIN-28A in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.
Specifications

Specifications

LIN-28A
6D1F9
Unconjugated
10mM Phosphate (pH 7.4) with 0.03% Sodium Azide
Purified
LIN28A
Purified recombinant fragment of LIN28 (aa93-209) expressed in E. Coli (NP_078950).
23 kDa
0.1 ml
Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers, Stem Cells
Primary
79727
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
1.0 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunocytochemistry 1:200 - 1:1000
Q9H9Z2
CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only