Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lipid A, Clone: 26-5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP150094

Catalog No. NBP150094


Add to cart

Description

Description

Lipid A Monoclonal antibody specifically detects Lipid A in Bacteria samples. It is validated for ELISA.
Specifications

Specifications

Lipid A
26-5
Unconjugated
PBS, containing 0.1% bovine serum albumin. with 0.02% Sodium Azide
Mouse
IgG2b
1 ml
Store at 4C. Do not freeze.
Monoclonal
Bacteria
ELISA
0.1 mg/mL
ELISA 1:100-1:2000
Purified
The antibody reacts with the Lipid A antigen.
Protein A or G purified
RUO
Primary
The antibody is reactive with Lipid A using ELISA. Lipid A is part of LPS (lipopolysaccaride) present in gram-negative bacteria.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only