Learn More
anti-Ly-49l, Clone: YLI-90, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128086
Catalog No. NBP128086
Add to cart
Description
Description
Ly-49l Monoclonal antibody specifically detects Ly-49l in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Ly-49l
YLI-90
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
Killer cell lectin like receptor subfamily A member 9, Klra9, Ly49I receptor, LY49I1
Mouse
IgG1 Kappa
0.5 mg
Store at 4C. Do not freeze.
Monoclonal
B6 allele of the Ly49I receptor
Flow Cytometry
0.5 mg/mL
Flow Cytometry <= 1 ug/10^6 cells
Purified
Klra9
CHO-K1 cells transfected with B6 allele of Ly49I gene
Protein A or G purified
RUO
Primary
16640
Mouse