Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lymphotoxin-alpha/TNF-beta, DyLight 755, Clone: 15A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP149428IR

Catalog No. NBP149428IR


Add to cart

Description

Description

Lymphotoxin-alpha/TNF-beta Monoclonal antibody specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Lymphotoxin-alpha/TNF-beta
15A3
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
NP_001153212
LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1
Mouse
IgG2b Kappa
0.1 ml
Adaptive Immunity, Apoptosis, Asthma, Cytokine Research, Immunology, Innate Immunity
Primary
4049
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
LTA
Recombinant human LTA (35-205aa) purified from E. coli.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human LTA (35-205aa)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only