Learn More
MAP2 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: 5H11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192711V
Catalog No. NBP192711V
Add to cart
Description
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
MAP2
5H11
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
MAP2
Full length purified bovine MAP2 protein. Epitope maps to 1057-1507 of the human sequence.
280 kDa
0.1 ml
Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Phospho Specific, Stem Cell Markers
Primary
4133
Immunocytochemistry, Immunofluorescence, Western Blot
1mg/mL
Western Blot, Immunocytochemistry/Immunofluorescence
P11137
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, microtubule-associated protein 2
Mouse
IgG2b
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat