Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Mouse anti-Human, Mouse, Rat, FITC, Clone: 5H11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192711F

Catalog No. NBP192711F


Add to cart

Description

Description

MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

MAP2
5H11
FITC
PBS with 0.05% Sodium Azide
Purified
MAP2
Full length purified bovine MAP2 protein. Epitope maps to 1057-1507 of the human sequence.
280 kDa
0.1 ml
Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Phospho Specific, Stem Cell Markers
Primary
4133
Immunocytochemistry, Immunofluorescence
1mg/mL
Immunocytochemistry/Immunofluorescence
P11137
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, microtubule-associated protein 2
Mouse
IgG2b
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only