Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-MAVS, DyLight 488, Clone: 58N3E1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222378G
Description
MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Western Blot.Specifications
MAVS | |
Monoclonal | |
DyLight 488 | |
AAH44952.1 | |
MAVS | |
Within the range of amino acids 93-298 of human MAVS (isoform 1) were used as the immunogen for this antibody. | |
0.1 mL | |
Immune System Diseases, Immunology, Protein Kinase | |
57506 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot | |
58N3E1 | |
Western Blot | |
CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only