Learn More
anti-MBD1, Clone: 100B272.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233221
Catalog No. NBP233221
Add to cart
Description
Description
MBD1 Monoclonal antibody specifically detects MBD1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.
Specifications
Specifications
MBD1
100B272.1
Unconjugated
PBS - BSA free with No Preservative
Purified
MBD1
This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 391-405 (SESEDGAGSPPPYRR) of human MBD1; GenBank no ref NP_056671.2
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
There are many isoforms of human MBD1 ranging from 503 (~ 55kD) to 655 (~ 71kD) amino acids in length. Researchers are encouraged to use the provided immunogen sequence to determine if this antibody is suitable for testing a specific isoform of interest.
Western Blot, ChIP assay, Immunoprecipitation
0.5 mg/mL
Western Blot 2-4 ug/ml, Chromatin Immunoprecipitation 1:10-1:500, Immunoprecipitation
Q9UIS9
CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
Mouse
IgG1
0.1 mg
Chromatin Research
Primary
4152
Human, Mouse