Learn More
anti-MCAM/CD146, Biotin, Clone: c264, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128333
Catalog No. NBP128333
Add to cart
Description
Description
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Chicken, Turkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
Specifications
MCAM/CD146
c264
Biotin
PBS with 0.05% Sodium Azide
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin
Mouse
IgG2b Kappa
0.25 mg
Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
4162
Chicken, Turkey
Flow Cytometry, Immunocytochemistry, Immunoprecipitation
0.5mg/mL
Flow Cytometry 1 ug/10 cells, Immunoprecipitation 1:10-1:500, Immunocytochemistry
Purified
MCAM
Mixture of E15 and E16 thymocytes from H.B15, H.B19, and H.B21 chicken embryos
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Chicken MCAM, Mr 98 kDa (protein core approx. 84 kDa)