Learn More
anti-mCherry, DyLight 350, Clone: 1C51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196752UV
Catalog No. NBP196752UV
Add to cart
Description
Description
mCherry Monoclonal antibody specifically detects mCherry in All Species samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
mCherry
1C51
Western Blot, Immunohistochemistry
Purified
Mouse
IgG2a
Protein G purified
RUO
Primary
Does not cross react with GFP.
Immunohistochemistry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
This antibody is raised against recombinant full-length mCherry with His tag purified from E. coli.
28 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
All Species