Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-mCherry, FITC, Clone: 1C51, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196752F

Catalog No. NBP196752F


Add to cart

Description

Description

mCherry Monoclonal antibody specifically detects mCherry in All Species samples. It is validated for Immunohistochemistry.
Specifications

Specifications

mCherry
1C51
FITC
PBS with 0.05% Sodium Azide
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
This antibody is raised against recombinant full-length mCherry with His tag purified from E. coli.
28 kDa
0.1 ml
Cellular Markers, Epitope Tags
Primary
Does not cross react with GFP.
Immunohistochemistry
1mg/mL
Immunohistochemistry
Purified
Mouse
IgG2a
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
All Species
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only