Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Meta-Pneumovirus, DyLight 405, Clone: HMPV123, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP121631V
Description
Meta-Pneumovirus Monoclonal antibody specifically detects Meta-Pneumovirus in Virus samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Meta-Pneumovirus | |
HMPV123 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
Purified | |
GLYG | |
Sub-type A hMPV virus isolate NCL03-4/145 | |
Protein G purified | |
RUO | |
Primary | |
2799942 | |
Primate, Virus |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
hMPV, metapneumovirus | |
Mouse | |
IgG2a | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
NBP1-21631 recognises the nucleoprotein of all sub-types (A1, A2, B1 and B2) of hMPV. NBP1-21631 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only