Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I Antibody (F21-2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128305
Description
MHC Class I Monoclonal specifically detects MHC Class I in Avian, Chicken, Turkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Electron Microscopy.Specifications
MHC Class I | |
Monoclonal | |
0.5mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
HLA-E | |
A monomorphic epitope on avian MHC Class I molecules (Mr approx. 68 kDa) | |
0.25 mg | |
Immunology, Major Histocompatibility Complexes | |
3133 | |
Avian, Chicken, Turkey | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
F21-2 | |
FITC | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Chicken/Turkey MHC Class I | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction