Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-MHC Class II, DyLight 680, Clone: HIS19, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200462FR
Description
MHC Class II Monoclonal specifically detects MHC Class II in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
MHC Class II | |
Monoclonal | |
DyLight 680 | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Rat | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
HIS19 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
HLA-DMA | |
The immunogen for this antibody is MHC Class II. | |
0.1 mL | |
Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
3108 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only