Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MMP-1, Clone: 3B6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142544

Catalog No. NBP142544


Add to cart

Description

Description

MMP-1 Monoclonal antibody specifically detects MMP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

MMP-1
3B6
Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin
Supernatant
MMP1
The antibody reacts with the MMP-1 antigen.
Tissue culture supernatant
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This monoclonal antibody binds to human MMP-1. Reactivity in other species has not been determined. The antibody was tested for cross-reactivity with MMP-2, MMP-3, and MMP-9 and did not cross-react. The 3B6 monoclonal antibody was generated from mice immunized with a 12 mer-peptide derived from the sequence of human MMP-1. The amino acid sequence of this peptide was: CSSFGFPRTVKH (Murray et al., 1998a and 1998b).
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Hybridoma culture supernatant with 0.05% Sodium Azide
CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP-1
Mouse
IgG1
0.2 ml
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix
Primary
4312
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only