Learn More
anti-MMP-1, Clone: SB12e, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP128604
Catalog No. NBP128604
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MMP-1
SB12e
Unconjugated
BBS (pH 8.2) with No Preservative
Purified
MMP1
Full-length recombinant MMP-1.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human MMP-1; Does not cross react to human MMP-2, MMP-3 or MMP-9
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 1:100-1:2000, ELISA 1:3000-1:8000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen < = 2 ug/ml
P03956
CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP-1
Mouse
IgG2b Kappa
0.1 mg
Angiogenesis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix
Primary
4312
Human