Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MMP-3, Clone: SB14d, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128615

Catalog No. NBP128615

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

MMP-3
SB14d
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1
Mouse
IgG2a
0.1 mg
Angiogenesis, Cancer
Primary
4314
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
0.5 mg/mL
ELISA 1:3000-1:8000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin < = 2 ug/ml, Immunohistochemistry-Frozen < = 2 ug/ml
Purified
MMP3
Recombinant MMP-3
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human MMP-3; Does not cross react to human MMP-1, MMP-2 or MMP-9
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only