Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Moesin, APC, Clone: SPM562, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234798APC

Catalog No. NBP234798J


Add to cart

Description

Description

Moesin Monoclonal antibody specifically detects Moesin in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

Moesin
SPM562
Flow Cytometry
Purified
MSN
Recombinant human Moesin protein
78 kDa
0.1 ml
Cytoskeleton Markers, Stem Cell Markers
Primary
4478
Human, Rat
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
Membrane-organizing extension spike protein, moesin
Mouse
IgG1 Kappa
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only