Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Monocyte/Granulocyte, APC, Clone: 74-22-15, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128241APC

Catalog No. NBP128241J


Add to cart

Description

Description

Monocyte/Granulocyte Monoclonal antibody specifically detects Monocyte/Granulocyte in Porcine samples. It is validated for Flow Cytometry.
Specifications

Specifications

Monocyte/Granulocyte
74-22-15
Flow Cytometry
Purified
IL8
Fresh dd miniature swine thymocytes
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Porcine SWC3a
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Mouse
IgG1 Kappa
0.25 ml
Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
3576
Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only