Learn More
anti-Monocyte/Granulocyte, DyLight 405, Clone: 74-22-15, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128241V
Catalog No. NBP128241V
Add to cart
Description
Description
Monocyte/Granulocyte Monoclonal antibody specifically detects Monocyte/Granulocyte in Porcine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Monocyte/Granulocyte
74-22-15
Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
IL8
Fresh dd miniature swine thymocytes
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Porcine SWC3a
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Mouse
IgG1 Kappa
0.25 ml
Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
3576
Porcine