Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MRP3, Clone: M3II-9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142347

Catalog No. NBP142347


Add to cart

Description

Description

MRP3 Monoclonal antibody specifically detects MRP3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

MRP3
M3II-9
Unconjugated
Serum - free culture supernatant with 0.7% BSA with 0.1% Sodium Azide
ABC31, ATP-binding cassette sub-family C member 3, ATP-binding cassette, sub-family C (CFTR/MRP), member 3, canalicular multispecific organic anion transporter 2, canicular multispecific organic anion transporter, cMOAT2, EC 3.6.3, EC 3.6.3.44, EC 6.3.2.4, MLP2DKFZp686E22157, MRP3MOAT-DEST90757, multidrug resistance associated protein, Multidrug resistance-associated protein 3, Multi-specific organic anion transporter D
Mouse
IgG1
0.2 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
M3II-9 reacts with an internal epitope of MRP3, a 190-200 kD transmembrane protein that is closely related to the multidrug resistance protein MRP1. M3II-9 was raised against a bacterial fusion protein of MRP3, containing amino acids 830-949 of the protein. M3II-9 does not cross-react with the human MDR1, MRP1, MRP2 or MRP5 gene products
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
0.25 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Supernatant
ABCC3
The antibody reacts with the MRP3 antigen.
Tissue culture supernatant
RUO
Primary
8714
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only