Learn More
anti-MS4A1/CD20, Alexa Fluor 488, Clone: MEM-97, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP144634AF488
Catalog No. NBP144634X
Add to cart
Description
Description
CD20 Monoclonal specifically detects CD20 in Human, Porcine, Bovine samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD20
MEM-97
Flow Cytometry
P11836
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Mouse
IgG1
0.1 ml
Adaptive Immunity, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers
Primary
931
Human, Porcine, Bovine
Flow Cytometry
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
MS4A1
Raji human Burkitt's lymphoma cell line (NM_021950.3).
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The antibody MEM-97 reacts with CD20 (Bp35), a 33-37 kDa non-glycosylated membrane receptor with four transmembrane domains, expressed on B lymphocytes (it is lost on plasma cells), follicular dendritic cells, and at low levels on peripheral blood T lymphocytes. HLDA V; WS Code B CD20.9