Learn More
anti-MSH2, Clone: 1184CT1.3.2, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232300
Catalog No. NBP232300
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MSH2
1184CT1.3.2
Unconjugated
PBS with 0.09% Sodium Azide
Purified
MSH2
Purified His-tagged MSH2 protein was used to produced this monoclonal antibody.
104.743 kDa
400 ul
Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors
Primary
4436
Western Blot
0.5 mg/mL
Western Blot 1:2000
P43246
COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human