Learn More
anti-MUC2, DyLight 350, Clone: CCP58, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233157UV
Catalog No. NBP233157UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MUC2
CCP58
ELISA
Purified
MUC2
A synthetic peptide of 29 amino acids, KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein, coupled to KLH.
520 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes a single glycoprotein of 520kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium. MUC2 is specifically expressed in goblet cells of the small intestine & colon; in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of the ovary.
ELISA
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Intestinal mucin-2, MLP, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, mucin-like protein, SMUC
Mouse
IgG1 Kappa
Protein A or G purified
RUO
Primary
4583
Human