Learn More
anti-Mucin 5AC, Clone: 2-12M1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142204
Catalog No. NBP142204
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MUC5AC Monoclonal antibody specifically detects MUC5AC in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Mucin 5AC
2-12M1
Unconjugated
0.1 mg/ml PBS with 0.05% Sodium Azide
gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin
Mouse
IgG1
0.1 mg
Extracellular Matrix
Primary
4586
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
0.1 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
MUC5AC
The antibody reacts with the MUC5AC antigen.
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
2-12M1 reacts with peptide core of gastric mucin (MUC5AC). Source: A BALB/c mouse was immunized with mucin isolated from an ovarian cyst fluid (pure endocervical type according to the Fenoglio's classification). Splenocytes were fused with mouse myeloma SP2/0 cells.