Learn More
anti-MyD88, Clone: 603E10.06, Novus Biologicals™
Manufacturer:  Novus Biologicals DDX1401P100
Catalog No. DDX1401P100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MyD88
603E10.06
Unconjugated
Tris-NaCl (pH 8.0) with No Preservative
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Mouse
IgG1
0.1 mg
Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary
4615
Flow Cytometry, Immunocytochemistry, Immunofluorescence
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Ion Exchange Chromatography
MYD88
Myd88 was used as immunogen.
Ion exchange chromatography
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Canine, Human