Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-Myeloperoxidase/MPO, Clone: 9B12G7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147430

Catalog No. NBP147430



Description

Description

Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Myeloperoxidase/MPO
9B12G7
Western Blot 1:500 - 1:2000, ELISA 1:10000
P05164
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Mouse
IgG1
0.1 ml
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Primary
4353
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
Ascites
MPO
Purified recombinant fragment of MPO(aa1-193) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only