Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MYST1, Clone: 8C4C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128879

Catalog No. NBP128879


Add to cart

Description

Description

MYST1 Monoclonal antibody specifically detects MYST1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

MYST1
8C4C4
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Q9H7Z6
FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1
Mouse
IgG2b
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
Ascites
KAT8
Purified recombinant fragment of human MYST1 expressed in E. Coli.
53 kDa
0.1 ml
Chromatin Research, DNA Repair
Primary
84148
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only