Learn More
anti-Nav1.8, Alexa Fluor 488, Clone: S134-12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147615AF488
Catalog No. NBP147615X
Add to cart
Description
Description
Nav1.8 Monoclonal antibody specifically detects Nav1.8 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Nav1.8
S134-12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
SCN10A
Fusion protein amino acids 1724-1956 of rat Nav1.8
Protein G purified
RUO
Primary
6336
Human, Monkey, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
hPN3, Nav1.8, PN3, sodium channel protein type 10 subunit alpha, sodium channel protein type X subunit alpha, sodium channel, voltage-gated, type X, alpha subunit, voltage-gated, type X, alpha polypeptide
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 220kDa. No cross reactivity against other Nav channels.