Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NCOA3/AIB1, Clone: 2C11B12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147424

Catalog No. NBP147424


Add to cart

Description

Description

NCOA3/AIB1 Monoclonal antibody specifically detects NCOA3/AIB1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

NCOA3/AIB1
2C11B12
Western Blot 1:500 - 1:2000, ELISA 1:10000
Q9Y6Q9
ACTRCBP-interacting protein, AIB1AIB-1, Amplified in breast cancer 1 protein, BHLHE42, bHLHe42p/CIP, CAGH16, Class E basic helix-loop-helix protein 42, CTG26, KAT13B, MGC141848, NCoA-3, nuclear receptor coactivator 3, pCIP RAC-3, RAC3pCIP, Receptor-associated coactivator 3, SRC3, SRC-3EC 2.3.1.48, Steroid receptor coactivator protein 3, Thyroid hormone receptor activator molecule 1, TNRC16, TRAM1, TRAM-1TNRC14
Mouse
IgG1
0.1 ml
Breast Cancer, Cancer, Chromatin Research, Transcription Factors and Regulators
Primary
8202
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
Ascites
NCOA3
Purified recombinant fragment of AIB1(aa1-200) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only