Learn More
anti-Nectin-2/CD112, Clone: R2-525, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142529
Catalog No. NBP142529
Add to cart
Description
Description
Nectin-2/CD112 Monoclonal antibody specifically detects Nectin-2/CD112 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
Specifications
Nectin-2/CD112
R2-525
Unconjugated
Phosphate buffered saline pH 7.4 with 0.09% Sodium Azide
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2
Mouse
IgG1
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes CD112 Poliovirus Receptor Related 2 (PRR2), a new surface molecule, which has been reported to be homologous to the poliovirus receptor (CD155). Studies demonstrate that PRR2 expression is restricted to the myelo-monocytic and megakaryocytic lineages. PRR2 expression has also been detected on haematopoeitic progenitors expressing CD34.
CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
1.0 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Purified
NECTIN2
The antibody reacts with the CD112 antigen.
Protein A or G purified
RUO
Primary
5819
Rat