Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurogenin-1 Mouse anti-Human, Clone: OTI3F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147871

Catalog No. NBP147871


Add to cart

Description

Description

Neurogenin-1 Monoclonal antibody specifically detects Neurogenin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).
Specifications

Specifications

Neurogenin-1
OTI3F9
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1
Mouse
IgG1
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens neurogenin 1 (NEUROG1).
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1:500-1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 2-4 ug/mg, Immunohistochemistry-Paraffin 1:50
Affinity Purified
NEUROG1
Full length human recombinant protein of human NEUROG1 (NP_006152) produced in HEK293T cell.
25.7 kDa
0.1 ml
Neuroscience
Primary
4762
Human, Monkey, Primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only