Learn More
NFkB p105/p50 Antibody (2J10D7), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB10056583UV
Catalog No. NB10056583U
Add to Cart
Description
Description
NFkB p105/p50 Monoclonal specifically detects NFkB p105/p50 in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
Specifications
NFkB p105/p50
Monoclonal
DyLight 350
P25942
NFKB1
A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen.
0.1 mL
Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
4790
Human, Rat, Rabbit
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
2J10D7
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50
Mouse
Protein G purified
RUO
Primary
Cross reacts with Human. Predicted to react with Mouse and Rat which are 86% homologous with the immunogen.
Store at 4C in the dark.
IgG1 κ
Product Suggestions
Product Suggestions