Learn More
anti-NFkB1/NFkB p105, Clone: 1298CT792.105.117.133, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232738
Catalog No. NBP232738
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
NFkB1/NFkB p105
1298CT792.105.117.133
Unconjugated
PBS with 0.09% Sodium Azide
Purified
NFKB1
This antibody is generated from a mouse immunized with a recombinant protein from human NFKB1.
105.356 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 1:1000, Flow Cytometry 1:25, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:25
P19838, P19838
DNA-binding factor KBF1, EBP-1, NF-kappa-B, NF-kB1, NFKB1, NFKB-p105, Nuclear factor NF-kappa-B p105 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105
Mouse
IgG1 Kappa
Protein G purified
RUO
Primary
4790