Learn More
anti-NHE3/SLC9A3, DyLight 650, Clone: 19F5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11061586C
Catalog No. NB11061586C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
NHE3/SLC9A3 Monoclonal antibody specifically detects NHE3/SLC9A3 in Rat, Rabbit, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
NHE3/SLC9A3
19F5
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
SLC9A3
Fusion protein representing the C-terminal 131 amino acids of rabbit NHE3. [UniProt# P26432]
93 kDa
0.1 ml
Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary
6550
Rat, Rabbit, Human (Negative)
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is specific for NHE3. It does not show any cross-reactivity with any other NHE isoforms.