Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse NKp46/NCR1, Fluorescein, Polyclonal, R&D Systems™

Manufacturer:  R&D Systems FAB2225F025

 View more versions of this product

Catalog No. FAB2225F025


Add to cart

Description

Description

Host Species: Goat
Species Reactivity:Mouse
Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse NKp46/NCR1, Glu22-Asn255, Accession No. Q8C567

Applications:

Flow Cytometry

Specifications

Specifications

NKp46/NCR1
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
NCR1
Mouse myeloma cell line NS0-derived recombinant mouse NKp46/NCR1, Glu22-Asn255, Accession # Q8C567
Antigen Affinity-purified
RUO
Primary
9437
Mouse
Flow Cytometry
Flow Cytometry 10 uL/10^6 cells
Q8C567
CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Goat
IgG
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Polyclonal
Detects mouse NKp46/NCR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human (rh) NKp46 and less than 1% cross-reactivity with rhNKp80 and rhNKp30 is observed.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit