Learn More
NKX3.1 Antibody (0361), Alexa Fluor™ 700, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB1001828AF700
Catalog No. NB1001828Z
Add to Cart
Description
Description
NKX3.1 Monoclonal specifically detects NKX3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
NKX3.1
Monoclonal
Alexa Fluor 700
Q99801
NKX3-1
Full-length human Nkx3.1 produced in bacteria as a hexahistidine-tagged protein. [UniProt# Q99801]
0.1 mL
Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators
4824
Mouse, Rat
Purified
KnockDown, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
361
Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A
Mouse
Protein G purified
RUO
Primary
Mouse and Rat. Predicted to work in human based on 100% sequence homology.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions