Learn More
anti-NPIP, Clone: 39C7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP130038
Catalog No. NBP130038
Add to cart
Description
Description
NPIP Monoclonal antibody specifically detects NPIP in Human, Mouse, Rat, Yeast samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunofluorescence.
Specifications
Specifications
NPIP
39C7
Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunofluorescence 1:10 - 1:500
Supernatant
NPIP
Yeast nuclear preparations
Concentrated Supernatant
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Reacts with a nuclear pore complex protein.
Immunocytochemistry, Immunofluorescence
Unconjugated
PBS with 10mM Sodium Azide
FLJ42525, FLJ44848, nuclear pore complex interacting protein, nuclear pore complex-interacting protein
Mouse
IgG1
0.1 ml
Neuroscience
Primary
9284
Human, Mouse, Rat, Yeast