Learn More
anti-NQO-1, Clone: MM0641-1A80, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232728
Catalog No. NBP232728
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
NQO-1
MM0641-1A80
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
NQO1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human NQO1
0.1 mg
Cancer, Cellular Markers, Lipid and Metabolism, Signal Transduction
Primary
1728
Human
Western Blot, Immunocytochemistry, Immunofluorescence
LYOPH
Western Blot 1:1000-2000, Immunocytochemistry/Immunofluorescence 1:50-200
Q3B792
Azoreductase, DHQU, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), dioxin-inducible 1, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1diaphorase-4, Phylloquinone reductase, QR1NMORI, Quinone reductase 1
Mouse
Protein G purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.