Learn More
anti-NRAS, Clone: 822CT17.5.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232298
Catalog No. NBP232298
Add to cart
Description
Description
NRAS Monoclonal antibody specifically detects NRAS in Human, Rat samples. It is validated for Western Blot.
Specifications
Specifications
NRAS
822CT17.5.1
Unconjugated
PBS with 0.09% Sodium Azide
Purified
NRAS
This NRAS antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 70-101 amino acids from the N-terminal region of human NRAS.
21.229 kDa
400 ul
Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway
Primary
4893
Western Blot
0.5 mg/mL
Western Blot 1:1000
P01111
ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Rat