Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-NT-4, Clone: 2E8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147897

Catalog No. NBP147897



Description

Description

NT-4 Monoclonal antibody specifically detects NT-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

NT-4
OTI2E8
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
GLC10, GLC1O, neurotrophic factor 4, neurotrophic factor 5, neurotrophin 4, neurotrophin 5 (neurotrophin 4/5), neurotrophin-4, neurotrophin-5, Neutrophic factor 4, NT4, NT-4, NT-4/5, NT-5, NTF5NT5
Mouse
IgG1
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens neurotrophin 4 (NTF4).
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Western Blot 1:5000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50
Affinity Purified
NTF4
Human recombinant protein fragment corresponding to amino acids 81-210 human NT4 (NP_006170) produced in E.coli.
22.2 kDa
0.1 ml
Cancer, Cytokine Research, Immunology, Neuroscience
Primary
4909
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only