Learn More
anti-NTB-A/SLAMF6/CD352, DyLight 405, Clone: 13G3-19D, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143344V
Catalog No. NBP143344V
Add to cart
Description
Description
NTB-A/SLAMF6/CD352 Monoclonal antibody specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
NTB-A/SLAMF6/CD352
13G3-19D
Western Blot, Flow Cytometry
Affinity Purified
SLAMF6
The immunogen for this antibody was Ly108.
Protein A or G purified
RUO
Primary
114836
Flow Cytometry, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6
Mouse
IgG2a Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse