Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORF73/HHV8 Antibody (4C11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP130176
Description
ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ORF73/HHV8 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 | |
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ORF73/HHV8 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin) | |
4C11 | |
Unconjugated | |
O40947 | |
HHV8GP78 | |
Recombinant human ORF73/HHV8 (122-329aa) purified from E. coli (AAB62657). | |
0.1 mL | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
935114 | |
Virus | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction