Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ornithine Decarboxylase, DyLight 488, Clone: SPM565, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234800G

Catalog No. NBP234800G


Add to cart

Description

Description

Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Ornithine Decarboxylase
SPM565
Western Blot, Flow Cytometry, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
ODC1
Recombinant human ODC-1 protein
53 kDa
0.1 ml
Cell Cycle and Replication
Primary
4953
Human, Rat
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1
Mouse
IgG1 Kappa
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Recognizes a 53kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only