Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-p57 Kip2, Clone: 3E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151675

Catalog No. NBP151675


Add to cart

Description

Description

p57 Kip2 Monoclonal antibody specifically detects p57 Kip2 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

p57 Kip2
3000
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
CDKN1C
Purified recombinant fragment of human CDKN1C expressed in E. Coli.
57 kDa
0.1 ml
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary
1028
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
Beckwith-Wiedemann syndrome, BWS, cyclin-dependent kinase inhibitor 1C, cyclin-dependent kinase inhibitor 1C (p57, Kip2), Cyclin-dependent kinase inhibitor p57, KIP2BWCR, p57, p57Kip2, WBS
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only