Learn More
anti-Palladin, Alexa Fluor 405, Clone: 1E6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP125959AF405
Catalog No. NBP125959W
Add to cart
Description
Description
Palladin Monoclonal antibody specifically detects Palladin in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Palladin
1000000
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Q8WX93
CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat, Chicken
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
PALLD
Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts.
85 kDa
0.1 ml
Cellular Markers, Cytoskeleton Markers, Signal Transduction
Primary
23022